Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCCTTCCCAAGGTCCCATTGAT[C/T]GGAAGCCTCCCCGAAGCCCGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137241 MIM: 601061 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIPR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIPR - gastric inhibitory polypeptide receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QPCTL - glutaminyl-peptide cyclotransferase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163377.1 | 519 | Silent Mutation | ATC,ATT | I,I 73 | NP_001156849.1 | |
NM_017659.3 | 519 | Silent Mutation | ATC,ATT | I,I 73 | NP_060129.2 | |
XM_011527048.2 | 519 | Silent Mutation | ATC,ATT | I,I 73 | XP_011525350.1 | |
XM_017026900.1 | 519 | Silent Mutation | ATC,ATT | I,I 73 | XP_016882389.1 |
SNRPD2 - small nuclear ribonucleoprotein D2 polypeptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |