Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATTGACTTTAAAATTAGGACCA[C/T]AGAGCTCGATGGCAAGAGAATTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165040 | ||||||||||||||||||||
Literature Links: |
RAB8A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB8A - RAB8A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005370.4 | 159 | Missense Mutation | ACA,ATA | T,I 50 | NP_005361.2 | |
XM_011528020.2 | 159 | Missense Mutation | ACA,ATA | T,I 50 | XP_011526322.1 |