Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGCCAGCGGGCGTTGACAGTG[A/G]CCAGCGTAAGGCTCAGGAACAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604350 MIM: 613771 | ||||||||||||||||||||
Literature Links: |
CCDC159 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC159 - coiled-coil domain containing 159 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB3D - RAB3D, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM205 - transmembrane protein 205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145416.1 | 294 | Missense Mutation | GCC,GTC | A,V 99 | NP_001138888.1 | |
NM_001321112.1 | 294 | Missense Mutation | GCC,GTC | A,V 99 | NP_001308041.1 | |
NM_001321113.1 | 294 | Missense Mutation | GCC,GTC | A,V 99 | NP_001308042.1 | |
NM_001321114.1 | 294 | Missense Mutation | GCC,GTC | A,V 99 | NP_001308043.1 | |
NM_033408.3 | 294 | Missense Mutation | GCC,GTC | A,V 99 | NP_212133.1 | |
NM_198536.2 | 294 | Missense Mutation | GCC,GTC | A,V 99 | NP_940938.1 | |
XM_005259899.2 | 294 | Missense Mutation | GCC,GTC | A,V 99 | XP_005259956.1 | |
XM_017026769.1 | 294 | Silent Mutation | GGC,GGT | G,G 92 | XP_016882258.1 |