Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGGCCTCACACTAGAGAGAAA[C/T]CTTTTGATTGTAAGGAATGTGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF440 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF440 - zinc finger protein 440 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF491 - zinc finger protein 491 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152356.3 | 530 | Missense Mutation | CCT,TCT | P,S 102 | NP_689569.2 | |
XM_005259730.3 | 530 | Missense Mutation | CCT,TCT | P,S 102 | XP_005259787.1 |