Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCGGCTCCTGGACAAAGACACAG[C/G]GGAGCTGCTGGGCGAGTGAGTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600944 MIM: 609458 MIM: 616850 | ||||||||||||||||||||
Literature Links: |
DHPS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHPS - deoxyhypusine synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206974.1 | 1013 | Intron | NP_001193903.1 | |||
NM_001930.3 | 1013 | Intron | NP_001921.1 | |||
NM_013406.2 | 1013 | Intron | NP_037538.1 | |||
XM_011527770.1 | 1013 | Intron | XP_011526072.1 | |||
XM_011527771.2 | 1013 | Intron | XP_011526073.1 |
MAN2B1 - mannosidase alpha class 2B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR83 - WD repeat domain 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099737.2 | 1013 | Missense Mutation | GCG,GGG | A,G 223 | NP_001093207.1 | |
NM_032332.3 | 1013 | Missense Mutation | GCG,GGG | A,G 223 | NP_115708.1 |
WDR83OS - WD repeat domain 83 opposite strand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |