Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCATCACTCCCTGCCTTTTACCC[C/T]GGCACTTCAGGTATCACTTCCACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604089 MIM: 604142 | ||||||||||||||||||||
Literature Links: |
HCST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCST - hematopoietic cell signal transducer | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007469.1 | 213 | Silent Mutation | CCC,CCT | P,P 33 | NP_001007470.1 | |
NM_014266.3 | 213 | Silent Mutation | CCC,CCT | P,P 33 | NP_055081.1 | |
XM_017026193.1 | 213 | Silent Mutation | CCC,CCT | P,P 33 | XP_016881682.1 |
NFKBID - NFKB inhibitor delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TYROBP - TYRO protein tyrosine kinase binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173514.1 | 213 | Intron | NP_001166985.1 | |||
NM_001173515.1 | 213 | Intron | NP_001166986.1 | |||
NM_003332.3 | 213 | Intron | NP_003323.1 | |||
NM_198125.2 | 213 | Intron | NP_937758.1 |