Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTGCCAAAGTCAAGCTGGCTC[A/G]GCACATCCTCACTGGTCGGGAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606495 | ||||||||||||||||||||
Literature Links: |
MARK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MARK4 - microtubule affinity regulating kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199867.1 | 561 | Missense Mutation | CAG,CGG | Q,R 77 | NP_001186796.1 | |
NM_031417.3 | 561 | Missense Mutation | CAG,CGG | Q,R 77 | NP_113605.2 | |
XM_006723307.3 | 561 | Intron | XP_006723370.1 |