Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACATGCTTTTGAAAGTACTGA[G/T]CGTGGCTGAAGGCTTTCCCACATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194551 | ||||||||||||||||||||
Literature Links: |
ZNF77 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF77 - zinc finger protein 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021217.2 | 1583 | Missense Mutation | GAT,GCT | D,A 476 | NP_067040.1 | |
XM_017027081.1 | 1583 | Missense Mutation | GAT,GCT | D,A 296 | XP_016882570.1 | |
XM_017027082.1 | 1583 | Missense Mutation | GAT,GCT | D,A 296 | XP_016882571.1 |