Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAAAGCGGACGCCCATCTTGAT[A/G]TTGGAGAACACGTGGGTGACCTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609099 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO27 - F-box protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178820.4 | 849 | Silent Mutation | AAC,AAT | N,N 243 | NP_849142.1 | |
XM_017026290.1 | 849 | Intron | XP_016881779.1 |