Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCCAGACTCATGAACAAGCTA[C/T]AGCCGGGCTCCGTCCCCAAGATCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605414 MIM: 602373 | ||||||||||||||||||||
Literature Links: |
ABCA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCA7 - ATP binding cassette subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNN2 - calponin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303499.1 | 589 | Nonsense Mutation | CAG,TAG | Q,* 68 | NP_001290428.1 | |
NM_001303501.1 | 589 | Nonsense Mutation | CAG,TAG | Q,* 68 | NP_001290430.1 | |
NM_004368.3 | 589 | Nonsense Mutation | CAG,TAG | Q,* 68 | NP_004359.1 | |
NM_201277.2 | 589 | Nonsense Mutation | CAG,TAG | Q,* 68 | NP_958434.1 |