Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACGGCTTGTCCGGGGGCAGCGA[C/G]AGCAGGCGGGTCGGTGAGAGGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604999 MIM: 600327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf81 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf81 - chromosome 19 open reading frame 81 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHANK1 - SH3 and multiple ankyrin repeat domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016148.3 | 6265 | Silent Mutation | CTC,CTG | L,L 2082 | NP_057232.2 | |
XM_006723233.3 | 6265 | Silent Mutation | CTC,CTG | L,L 2082 | XP_006723296.1 | |
XM_011527013.2 | 6265 | Silent Mutation | CTC,CTG | L,L 2090 | XP_011525315.1 | |
XM_011527014.2 | 6265 | Silent Mutation | CTC,CTG | L,L 2073 | XP_011525316.1 |
SYT3 - synaptotagmin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |