Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGTAGACCACGAAACACAGCT[C/T]CTGGCCCCAGTGTGTCATGGACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603159 MIM: 607959 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP3 - LDL receptor related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002333.3 | 1247 | Intron | NP_002324.2 | |||
XM_005258945.1 | 1247 | Intron | XP_005259002.1 |
SLC7A10 - solute carrier family 7 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019849.2 | 1247 | Missense Mutation | AAG,GAG | K,E 489 | NP_062823.1 | |
XM_006723284.2 | 1247 | Missense Mutation | AAG,GAG | K,E 410 | XP_006723347.1 | |
XM_011527119.1 | 1247 | Missense Mutation | AAG,GAG | K,E 626 | XP_011525421.1 | |
XM_011527120.1 | 1247 | Missense Mutation | AAG,GAG | K,E 599 | XP_011525422.1 |