Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAACAGGATCTTGCTCTGGGCGC[A/G]CAAGGGCTGGTCACGCTCCGCCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612685 MIM: 614770 MIM: 610850 | ||||||||||||||||||||
Literature Links: |
CAMSAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMSAP3 - calmodulin regulated spectrin associated protein family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6792 - microRNA 6792 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PET100 - PET100 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XAB2 - XPA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020196.2 | 2394 | Missense Mutation | CGC,TGC | R,C 790 | NP_064581.2 |