Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCACCTAGGGCACCATCGGGAG[G/T]CCAATGGGGTCATATGGAGACACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SIGLEC16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIGLEC16 - sialic acid binding Ig like lectin 16 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VRK3 - vaccinia related kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025778.1 | 1894 | Silent Mutation | GGA,GGC | G,G 419 | NP_001020949.1 | |
NM_001308420.1 | 1894 | Intron | NP_001295349.1 | |||
NM_016440.3 | 1894 | Silent Mutation | GGA,GGC | G,G 469 | NP_057524.3 | |
XM_005258971.3 | 1894 | Silent Mutation | GGA,GGC | G,G 469 | XP_005259028.1 | |
XM_005258972.3 | 1894 | Silent Mutation | GGA,GGC | G,G 419 | XP_005259029.1 | |
XM_006723237.3 | 1894 | Silent Mutation | GGA,GGC | G,G 469 | XP_006723300.1 | |
XM_006723238.3 | 1894 | Silent Mutation | GGA,GGC | G,G 419 | XP_006723301.1 | |
XM_011527023.2 | 1894 | Intron | XP_011525325.1 |