Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGGTCAGAGCCTGAGTCCTCG[C/T]CTTGTTGGTTCTGAATCTGACTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126391 MIM: 603602 | ||||||||||||||||||||
Literature Links: |
LIG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIG1 - DNA ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000234.2 | 2672 | Missense Mutation | GAC,GGC | D,G 908 | NP_000225.1 | |
NM_001289063.1 | 2672 | Missense Mutation | GAC,GGC | D,G 877 | NP_001275992.1 | |
NM_001289064.1 | 2672 | Missense Mutation | GAC,GGC | D,G 840 | NP_001275993.1 | |
NM_001320970.1 | 2672 | Missense Mutation | GAC,GGC | D,G 907 | NP_001307899.1 | |
NM_001320971.1 | 2672 | Missense Mutation | GAC,GGC | D,G 878 | NP_001307900.1 | |
XM_017026809.1 | 2672 | Missense Mutation | GAC,GGC | D,G 872 | XP_016882298.1 |
PLA2G4C - phospholipase A2 group IVC | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |