Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAAGGTGGCCAACTGCTGCTC[C/T]ATAGCAATAAGAAGGAAAGAACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608451 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETHE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETHE1 - ETHE1, persulfide dioxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320867.1 | Intron | NP_001307796.1 | ||||
NM_001320868.1 | Intron | NP_001307797.1 | ||||
NM_001320869.1 | Intron | NP_001307798.1 | ||||
NM_014297.4 | Intron | NP_055112.2 | ||||
XM_005258687.3 | Intron | XP_005258744.1 |
ZNF575 - zinc finger protein 575 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |