Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCGTCTTGGGGGCCTTGGCCTT[A/G]CCTGCTCCCAGCAGGGCCCCCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPERD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATSPERD - cation channel sperm associated auxiliary subunit delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUS3L - dihydrouridine synthase 3 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR22 - proline rich 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134316.1 | 906 | Silent Mutation | GGC,GGT | G,G 267 | NP_001127788.1 |