Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACAGGGCCAGGGGCACCAGCCA[C/T]GCATAGCAGTAGATGCTGATGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603934 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf52 - chromosome 19 open reading frame 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CARM1 - coactivator associated arginine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199141.1 | 589 | Intron | NP_954592.1 | |||
XM_005259708.4 | 589 | Intron | XP_005259765.1 | |||
XM_011527638.2 | 589 | Intron | XP_011525940.1 |
YIPF2 - Yip1 domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321439.1 | 589 | Silent Mutation | GCA,GCG | A,A 173 | NP_001308368.1 | |
NM_001321440.1 | 589 | Silent Mutation | GCA,GCG | A,A 134 | NP_001308369.1 | |
NM_024029.4 | 589 | Silent Mutation | GCA,GCG | A,A 173 | NP_076934.1 | |
XM_011528270.2 | 589 | Silent Mutation | GCA,GCG | A,A 173 | XP_011526572.1 | |
XM_011528271.2 | 589 | Silent Mutation | GCA,GCG | A,A 144 | XP_011526573.1 |