Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCTGGAGCACTCCTGCCCCATG[C/T]CCCACCTTGCCTGCTCTTCAGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607963 MIM: 606154 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBD3L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBD3L1 - methyl-CpG binding domain protein 3 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145208.2 | 444 | Missense Mutation | CCC,TCC | P,S 120 | NP_660209.2 |
MUC16 - mucin 16, cell surface associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |