Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCTCGGACCAGGACGGGCTGGC[C/T]CTGCCCCGCGGCCTCCTCCTCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605069 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKNK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKNK2 - MAP kinase interacting serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017572.3 | 1509 | Intron | NP_060042.2 | |||
NM_199054.2 | 1509 | Missense Mutation | AGC,GGC | S,G 422 | NP_951009.1 |