Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCCGTGTTTGAGAGCCTCGGG[C/T]GCCGTCCACTTGACGGGCAGCCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600038 MIM: 610362 | ||||||||||||||||||||
Literature Links: |
MATK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MATK - megakaryocyte-associated tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002378.3 | 1576 | Silent Mutation | GCA,GCG | A,A 393 | NP_002369.2 | |
NM_139354.2 | 1576 | Silent Mutation | GCA,GCG | A,A 351 | NP_647611.1 | |
NM_139355.2 | 1576 | Silent Mutation | GCA,GCG | A,A 392 | NP_647612.1 | |
XM_011528018.1 | 1576 | Silent Mutation | GCA,GCG | A,A 392 | XP_011526320.1 |
RAX2 - retina and anterior neural fold homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |