Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGACCACCATGCTGCTGCTGAC[C/T]GCCTCAGCCCAGCGCCTGTCACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613452 MIM: 603939 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPERG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATSPERG - cation channel sperm associated auxiliary subunit gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNK6 - potassium two pore domain channel subfamily K member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004823.1 | 1089 | Silent Mutation | ACC,ACT | T,T 139 | NP_004814.1 | |
XM_011527526.1 | 1089 | Silent Mutation | ACC,ACT | T,T 5 | XP_011525828.1 | |
XM_011527527.1 | 1089 | Intron | XP_011525829.1 | |||
XM_017027506.1 | 1089 | Intron | XP_016882995.1 |