Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCTCCACCATGCTGCTGATGA[A/G]CCTCGCGGCTGCTGACCTCCTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608841 MIM: 602779 MIM: 607777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPAMD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPAMD8 - C3 and PZP like, alpha-2-macroglobulin domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
F2RL3 - F2R like thrombin/trypsin receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003950.2 | 346 | Missense Mutation | NP_003941.2 | |||
XM_005260139.2 | 346 | Missense Mutation | AAC,AGC | N,S 5 | XP_005260196.1 |
SIN3B - SIN3 transcription regulator family member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |