Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGGGTGAGGGACACCAGATGAC[A/C]CTGTGCCTGGAGTGAGAAGGGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600137 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K10 - mitogen-activated protein kinase kinase kinase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002446.3 | Intron | NP_002437.2 | ||||
XM_006723222.2 | Intron | XP_006723285.1 | ||||
XM_006723223.3 | Intron | XP_006723286.1 | ||||
XM_011526981.2 | Intron | XP_011525283.1 | ||||
XM_011526982.2 | Intron | XP_011525284.1 |