Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTGCGCACATCTCGGAGGTGGC[A/G]GCTGATGGCCAGCAGGGTGCTGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609079 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL12 - F-box and leucine rich repeat protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316936.1 | 963 | Missense Mutation | CGC,TGC | R,C 192 | NP_001303865.1 | |
NM_001316937.1 | 963 | Missense Mutation | CGC,TGC | R,C 172 | NP_001303866.1 | |
NM_001316938.1 | 963 | Missense Mutation | CGC,TGC | R,C 172 | NP_001303867.1 | |
NM_001316939.1 | 963 | Missense Mutation | CGC,TGC | R,C 172 | NP_001303868.1 | |
NM_001316940.1 | 963 | Missense Mutation | CGC,TGC | R,C 172 | NP_001303869.1 | |
NM_001316941.1 | 963 | Missense Mutation | CGC,TGC | R,C 172 | NP_001303870.1 | |
NM_001316942.1 | 963 | Missense Mutation | CGC,TGC | R,C 172 | NP_001303871.1 | |
NM_017703.2 | 963 | Missense Mutation | CGC,TGC | R,C 225 | NP_060173.1 | |
XM_006722782.3 | 963 | Missense Mutation | CGC,TGC | R,C 183 | XP_006722845.2 | |
XM_017026912.1 | 963 | Missense Mutation | CGC,TGC | R,C 242 | XP_016882401.1 |