Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGACCCCCTCACACTCTCACCTT[C/T]ACCCGCAGCATCTCCTCGGGGATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600006 | ||||||||||||||||||||
Literature Links: |
DCAF15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAF15 - DDB1 and CUL4 associated factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFX1 - regulatory factor X1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002918.4 | 2984 | Silent Mutation | GTA,GTG | V,V 736 | NP_002909.4 | |
XM_011528165.2 | 2984 | Silent Mutation | GTA,GTG | V,V 846 | XP_011526467.1 | |
XM_011528166.2 | 2984 | Silent Mutation | GTA,GTG | V,V 846 | XP_011526468.1 | |
XM_011528167.2 | 2984 | Silent Mutation | GTA,GTG | V,V 846 | XP_011526469.1 | |
XM_011528168.2 | 2984 | Silent Mutation | GTA,GTG | V,V 748 | XP_011526470.1 | |
XM_011528169.2 | 2984 | Silent Mutation | GTA,GTG | V,V 737 | XP_011526471.1 | |
XM_011528170.2 | 2984 | Silent Mutation | GTA,GTG | V,V 737 | XP_011526472.1 |