Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCTGTCCCCCACAGGTGACCA[C/G]CAATGGCAGCATCGGGAGGGACCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604783 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLPTM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLPTM1 - CLPTM1, transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282175.1 | 125 | Missense Mutation | ACC,AGC | T,S 13 | NP_001269104.1 | |
NM_001282176.1 | 125 | UTR 5 | NP_001269105.1 | |||
NM_001294.3 | 125 | Missense Mutation | ACC,AGC | T,S 27 | NP_001285.1 |