Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAACTTTGCAGTTATGGCATC[C/T]CTAGGTAAGGCCCTCCTATTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604076 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF134 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF134 - zinc finger protein 134 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF530 - zinc finger protein 530 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321981.1 | 376 | Silent Mutation | TCC,TCT | S,S 19 | NP_001308910.1 | |
NM_020880.4 | 376 | Silent Mutation | TCC,TCT | S,S 52 | NP_065931.3 | |
XM_006723194.1 | 376 | Silent Mutation | TCC,TCT | S,S 52 | XP_006723257.1 | |
XM_011526919.2 | 376 | Silent Mutation | TCC,TCT | S,S 51 | XP_011525221.1 | |
XM_011526921.2 | 376 | Silent Mutation | TCC,TCT | S,S 19 | XP_011525223.1 |