Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAAAGTCACTGCCGCAATAGTG[G/T]AGTAGTCGGTGGGCGCGGCCGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTSL5 - ADAMTS like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213604.2 | 1399 | Silent Mutation | CTA,CTC | L,L 364 | NP_998769.2 | |
XM_005259549.3 | 1399 | Silent Mutation | CTA,CTC | L,L 374 | XP_005259606.1 | |
XM_011527961.1 | 1399 | Silent Mutation | CTA,CTC | L,L 378 | XP_011526263.1 | |
XM_011527963.1 | 1399 | Silent Mutation | CTA,CTC | L,L 133 | XP_011526265.1 | |
XM_017026744.1 | 1399 | Silent Mutation | CTA,CTC | L,L 368 | XP_016882233.1 | |
XM_017026745.1 | 1399 | Silent Mutation | CTA,CTC | L,L 364 | XP_016882234.1 |
REEP6 - receptor accessory protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |