Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGGGGGGTCTGCACTCACCATA[C/T]GCTGCTTCCCGACCCATGTCACTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603346 | ||||||||||||||||||||
Literature Links: |
NPAS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPAS1 - neuronal PAS domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321086.1 | 602 | Intron | NP_001308015.1 | |||
NM_002517.3 | 602 | Intron | NP_002508.2 | |||
XM_006723231.2 | 602 | Intron | XP_006723294.1 | |||
XM_011526997.1 | 602 | Intron | XP_011525299.1 | |||
XM_011526999.1 | 602 | Intron | XP_011525301.1 | |||
XM_011527001.1 | 602 | Intron | XP_011525303.1 | |||
XM_011527002.1 | 602 | Intron | XP_011525304.1 | |||
XM_011527006.1 | 602 | Intron | XP_011525308.1 | |||
XM_011527007.1 | 602 | Intron | XP_011525309.1 | |||
XM_017026841.1 | 602 | Intron | XP_016882330.1 | |||
XM_017026842.1 | 602 | Intron | XP_016882331.1 | |||
XM_017026843.1 | 602 | Intron | XP_016882332.1 | |||
XM_017026844.1 | 602 | Intron | XP_016882333.1 |
TMEM160 - transmembrane protein 160 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017854.1 | 602 | Silent Mutation | GCA,GCG | A,A 99 | NP_060324.1 | |
XM_017026917.1 | 602 | Silent Mutation | GCA,GCG | A,A 10 | XP_016882406.1 |