Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTCACAGATCCAGGGGTTGCC[A/G]GAGATGTCGAAGCCATCCCGCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611289 MIM: 613248 MIM: 608873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRG1 - leucine rich alpha-2-glycoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052972.2 | 902 | Silent Mutation | TCC,TCT | S,S 297 | NP_443204.1 |
PLIN5 - perilipin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA6B - semaphorin 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |