Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCATGGCCTTGACCAAGGAGCT[C/T]TCTGGCAGCTGGCGGAAGATGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116897 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEBPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEBPA - CCAAT/enhancer binding protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285829.1 | 1191 | Silent Mutation | GAA,GAG | E,E 228 | NP_001272758.1 | |
NM_001287424.1 | 1191 | Silent Mutation | GAA,GAG | E,E 382 | NP_001274353.1 | |
NM_001287435.1 | 1191 | Silent Mutation | GAA,GAG | E,E 333 | NP_001274364.1 | |
NM_004364.4 | 1191 | Silent Mutation | GAA,GAG | E,E 347 | NP_004355.2 |
CEBPA-AS1 - CEBPA antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |