Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGTGGATGACACCAGCAGTGA[A/G]GTGCTGGATGAGCTGTACCGCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606746 MIM: 615869 | ||||||||||||||||||||
Literature Links: |
MYDGF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYDGF - myeloid derived growth factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP8L1 - TNF alpha induced protein 8 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167942.1 | 944 | Silent Mutation | GAA,GAG | E,E 37 | NP_001161414.1 | |
NM_152362.2 | 944 | Silent Mutation | GAA,GAG | E,E 37 | NP_689575.2 | |
XM_005259487.3 | 944 | Silent Mutation | GAA,GAG | E,E 37 | XP_005259544.1 | |
XM_011527680.2 | 944 | Silent Mutation | GAA,GAG | E,E 37 | XP_011525982.1 |