Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATTACTCCAAGAACCTGGTGAC[A/G]GAGGACGTGATGCGGATGCTGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172400 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPI - glucose-6-phosphate isomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184722.1 | 240 | Intron | NP_001171651.1 | |||
NM_001289789.1 | 240 | Intron | NP_001276718.1 | |||
XM_005258764.2 | 240 | Silent Mutation | ACA,ACG | T,T 61 | XP_005258821.1 | |
XM_006723148.2 | 240 | Silent Mutation | ACA,ACG | T,T 61 | XP_006723211.1 | |
XM_011526754.2 | 240 | Silent Mutation | ACA,ACG | T,T 100 | XP_011525056.1 |