Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAGGCCGTGCTATTCCTGGGGG[A/G]TGTGCGCGGCCATCTGGGCCCTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 107266 MIM: 603820 MIM: 603821 | ||||||||||||||||||||
Literature Links: |
CD22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD22 - CD22 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FFAR1 - free fatty acid receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005303.2 | 386 | Missense Mutation | ATG,GTG | M,V 126 | NP_005294.1 |
FFAR3 - free fatty acid receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5196 - microRNA 5196 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |