Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTTCTTTCAAGTTTCTGAAATT[A/T]AAGTACTGAATGTTTTCCCACATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF823 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF823 - zinc finger protein 823 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080493.3 | 1929 | UTR 3 | NP_001073962.1 | |||
NM_001297610.1 | 1929 | UTR 3 | NP_001284539.1 | |||
NM_017507.1 | 1929 | UTR 3 | NP_059977.1 | |||
XM_017026941.1 | 1929 | UTR 3 | XP_016882430.1 | |||
XM_017026942.1 | 1929 | UTR 3 | XP_016882431.1 |