Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATTGGATGCCGGGAATCAACC[A/G]CCCTCTCAAAATATCTCCATTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612773 MIM: 608453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAM - basal cell adhesion molecule (Lutheran blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CBLC - Cbl proto-oncogene C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130852.1 | 1474 | Intron | NP_001124324.1 | |||
NM_012116.3 | 1474 | Intron | NP_036248.3 | |||
XM_005258696.3 | 1474 | Intron | XP_005258753.1 | |||
XM_011526688.2 | 1474 | UTR 3 | XP_011524990.1 | |||
XM_011526689.2 | 1474 | UTR 3 | XP_011524991.1 | |||
XM_011526690.2 | 1474 | Intron | XP_011524992.1 |