Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTCTTGCTGTGGGACAACCCAG[A/G]AGGAAGGACTCTTGCTGCCTCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZIM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZIM2 - zinc finger imprinted 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146326.1 | 1867 | Missense Mutation | CCT,TCT | P,S 442 | NP_001139798.1 | |
NM_001146327.1 | 1867 | Missense Mutation | CCT,TCT | P,S 442 | NP_001139799.1 | |
NM_015363.4 | 1867 | Missense Mutation | CCT,TCT | P,S 442 | NP_056178.3 |
ZIM2-AS1 - ZIM2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |