Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTTGAAATCACAACAAATTGAA[A/G]CTCCCAGAAATATAATAAGTATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR7A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR7A5 - olfactory receptor family 7 subfamily A member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017506.1 | 1199 | UTR 3 | NP_059976.1 | |||
XM_011527912.2 | 1199 | UTR 3 | XP_011526214.1 | |||
XM_011527913.2 | 1199 | UTR 3 | XP_011526215.1 | |||
XM_017026583.1 | 1199 | UTR 3 | XP_016882072.1 |
OR7C1 - olfactory receptor family 7 subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198944.1 | 1199 | Intron | NP_945182.1 | |||
XM_005259853.3 | 1199 | Intron | XP_005259910.1 | |||
XM_011527915.2 | 1199 | Intron | XP_011526217.1 |