Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGAAAGCGCCGGGTCTTCTCCA[C/T]GCCCTGCGTGTCCGAGCCCCAGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CATSPERD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CATSPERD - cation channel sperm associated auxiliary subunit delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUS3L - dihydrouridine synthase 3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161619.1 | 1799 | Missense Mutation | ATG,GTG | M,V 326 | NP_001155091.1 | |
NM_020175.2 | 1799 | Missense Mutation | ATG,GTG | M,V 568 | NP_064560.2 | |
XM_017027020.1 | 1799 | Missense Mutation | ATG,GTG | M,V 554 | XP_016882509.1 |
PRR22 - proline rich 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134316.1 | 1799 | Intron | NP_001127788.1 |