Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATGAACATGTCCACTGCATTGT[C/T]AGGGTTTGTTTCCATTATGAGTTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF432 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF432 - zinc finger protein 432 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322284.1 | 2580 | Silent Mutation | TAA,TGA | *,* 653 | NP_001309213.1 | |
NM_001322285.1 | 2580 | Silent Mutation | TAA,TGA | *,* 653 | NP_001309214.1 | |
NM_014650.3 | 2580 | Silent Mutation | TAA,TGA | *,* 653 | NP_055465.1 |
ZNF614 - zinc finger protein 614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |