Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTGGCTGTCGAAACAGGAGGCG[C/G]TGAGGAAGACAGGGGGAGCCGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF579 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF579 - zinc finger protein 579 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152600.2 | 1639 | Missense Mutation | ACC,AGC | T,S 529 | NP_689813.2 | |
XM_017026409.1 | 1639 | Missense Mutation | ACC,AGC | T,S 529 | XP_016881898.1 | |
XM_017026410.1 | 1639 | Missense Mutation | ACC,AGC | T,S 529 | XP_016881899.1 | |
XM_017026411.1 | 1639 | Missense Mutation | ACC,AGC | T,S 529 | XP_016881900.1 |