Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAACTGGAGCACTGAAAATCAC[A/G]TCTTGTGAGTTTTTCCATTATTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF566 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF566 - zinc finger protein 566 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145343.1 | 680 | Missense Mutation | CGT,TGT | R,C 112 | NP_001138815.1 | |
NM_001145344.1 | 680 | Missense Mutation | CGT,TGT | R,C 111 | NP_001138816.1 | |
NM_001145345.1 | 680 | Missense Mutation | CGT,TGT | R,C 111 | NP_001138817.1 | |
NM_001300970.1 | 680 | Missense Mutation | CGT,TGT | R,C 7 | NP_001287899.1 | |
NM_032838.4 | 680 | Missense Mutation | CGT,TGT | R,C 111 | NP_116227.1 | |
XM_005259354.2 | 680 | Missense Mutation | CGT,TGT | R,C 79 | XP_005259411.1 | |
XM_005259356.2 | 680 | Missense Mutation | CGT,TGT | R,C 7 | XP_005259413.1 | |
XM_006723447.2 | 680 | Missense Mutation | CGT,TGT | R,C 112 | XP_006723510.1 | |
XM_011527428.1 | 680 | Missense Mutation | CGT,TGT | R,C 7 | XP_011525730.1 | |
XM_017027399.1 | 680 | Missense Mutation | CGT,TGT | R,C 7 | XP_016882888.1 | |
XM_017027400.1 | 680 | Missense Mutation | CGT,TGT | R,C 7 | XP_016882889.1 |