Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCTGGTAGATGCTTCCTAAGTGG[A/C]GTTTTCAAGGTGGCCAGGCCCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMC6 - armadillo repeat containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199196.1 | Intron | NP_001186125.1 | ||||
NM_033415.3 | Intron | NP_219483.1 | ||||
XM_005260157.3 | Intron | XP_005260214.1 | ||||
XM_017027484.1 | Intron | XP_016882973.1 | ||||
XM_017027485.1 | Intron | XP_016882974.1 |
SUGP2 - SURP and G-patch domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |