Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGGTGGTGCCTGGAGGCCGTC[A/G]TGGAGGAGCAGATGGCCAGCGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD70 - CD70 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252.4 | 503 | Missense Mutation | ACG,ATG | T,M 118 | NP_001243.1 | |
XM_017027523.1 | 503 | Intron | XP_016883012.1 |