Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTTTCATTTACAGCCTTGGGAC[A/G]ATAATGCAGTCTGTGCAGACCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123837 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNE1 - cyclin E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001238.3 | 373 | Missense Mutation | AAT,GAT | N,D 64 | NP_001229.1 | |
NM_001322259.1 | 373 | Missense Mutation | AAT,GAT | N,D 64 | NP_001309188.1 | |
NM_001322261.1 | 373 | Missense Mutation | AAT,GAT | N,D 64 | NP_001309190.1 | |
NM_001322262.1 | 373 | Missense Mutation | AAT,GAT | N,D 49 | NP_001309191.1 | |
XM_011527440.2 | 373 | Missense Mutation | AAT,GAT | N,D 61 | XP_011525742.1 |