Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCATTGTCCCTCTACCCAAACT[C/T]TACCAGATTCATTTTTCGATACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF320 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF320 - zinc finger protein 320 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207333.2 | 3770 | Intron | NP_997216.2 | |||
XM_006723059.3 | 3770 | UTR 3 | XP_006723122.1 | |||
XM_017026393.1 | 3770 | UTR 3 | XP_016881882.1 | |||
XM_017026394.1 | 3770 | UTR 3 | XP_016881883.1 | |||
XM_017026395.1 | 3770 | UTR 3 | XP_016881884.1 | |||
XM_017026396.1 | 3770 | UTR 3 | XP_016881885.1 | |||
XM_017026397.1 | 3770 | UTR 3 | XP_016881886.1 | |||
XM_017026398.1 | 3770 | Intron | XP_016881887.1 | |||
XM_017026399.1 | 3770 | UTR 3 | XP_016881888.1 | |||
XM_017026400.1 | 3770 | UTR 3 | XP_016881889.1 |