Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGATGCAAAGGCCAGTGTAGAC[A/G]TCCTCAAAGGGGAAGGGTGCCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615357 MIM: 608348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5SL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5SL - ATP5S like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
B3GNT8 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198540.2 | 2775 | Silent Mutation | GAC,GAT | D,D 334 | NP_940942.1 | |
XM_011526934.2 | 2775 | Silent Mutation | GAC,GAT | D,D 334 | XP_011525236.1 |
BCKDHA - branched chain keto acid dehydrogenase E1, alpha polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000709.3 | 2775 | Intron | NP_000700.1 | |||
NM_001164783.1 | 2775 | Intron | NP_001158255.1 |