Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGTGCAGCTGCTCGGCGATCA[A/G]CAGGATCTCCTCTGAGGTAGGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164176 | ||||||||||||||||||||
Literature Links: |
POU2F2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POU2F2 - POU class 2 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207025.2 | 1380 | Silent Mutation | CTG,TTG | L,L 329 | NP_001193954.1 | |
NM_001207026.1 | 1380 | Silent Mutation | CTG,TTG | L,L 329 | NP_001193955.1 | |
NM_001247994.1 | 1380 | Silent Mutation | CTG,TTG | L,L 313 | NP_001234923.1 | |
NM_002698.4 | 1380 | Silent Mutation | CTG,TTG | L,L 313 | NP_002689.1 | |
XM_005259010.3 | 1380 | Silent Mutation | CTG,TTG | L,L 313 | XP_005259067.1 | |
XM_011527040.2 | 1380 | Silent Mutation | CTG,TTG | L,L 351 | XP_011525342.1 | |
XM_011527041.2 | 1380 | Silent Mutation | CTG,TTG | L,L 351 | XP_011525343.1 | |
XM_011527042.2 | 1380 | Silent Mutation | CTG,TTG | L,L 335 | XP_011525344.1 | |
XM_011527043.2 | 1380 | Silent Mutation | CTG,TTG | L,L 450 | XP_011525345.2 | |
XM_017026884.1 | 1380 | Silent Mutation | CTG,TTG | L,L 450 | XP_016882373.1 | |
XM_017026885.1 | 1380 | Silent Mutation | CTG,TTG | L,L 450 | XP_016882374.1 | |
XM_017026886.1 | 1380 | Silent Mutation | CTG,TTG | L,L 438 | XP_016882375.1 | |
XM_017026887.1 | 1380 | Silent Mutation | CTG,TTG | L,L 438 | XP_016882376.1 | |
XM_017026888.1 | 1380 | Silent Mutation | CTG,TTG | L,L 438 | XP_016882377.1 | |
XM_017026889.1 | 1380 | Silent Mutation | CTG,TTG | L,L 438 | XP_016882378.1 | |
XM_017026890.1 | 1380 | Silent Mutation | CTG,TTG | L,L 438 | XP_016882379.1 | |
XM_017026891.1 | 1380 | Silent Mutation | CTG,TTG | L,L 434 | XP_016882380.1 | |
XM_017026892.1 | 1380 | Silent Mutation | CTG,TTG | L,L 428 | XP_016882381.1 | |
XM_017026893.1 | 1380 | Silent Mutation | CTG,TTG | L,L 335 | XP_016882382.1 | |
XM_017026894.1 | 1380 | Silent Mutation | CTG,TTG | L,L 329 | XP_016882383.1 | |
XM_017026895.1 | 1380 | Silent Mutation | CTG,TTG | L,L 329 | XP_016882384.1 | |
XM_017026896.1 | 1380 | Silent Mutation | CTG,TTG | L,L 450 | XP_016882385.1 |