Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCATTGTGGCCCTGAAGGTTCT[C/T]TTCAAGTCGCAGATAGAGAAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603495 MIM: 604668 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AURKC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AURKC - aurora kinase C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015878.1 | 270 | Silent Mutation | CTC,CTT | L,L 74 | NP_001015878.1 | |
NM_001015879.1 | 270 | Silent Mutation | CTC,CTT | L,L 55 | NP_001015879.1 | |
NM_003160.2 | 270 | Silent Mutation | CTC,CTT | L,L 40 | NP_003151.2 |
ZNF264 - zinc finger protein 264 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF805 - zinc finger protein 805 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |